Learn More
System Biosciences LLC Exosome Cyto-Tracer, pCT-CD63-GFPpCT-CD63-GFP (pCMV, Exosome/Secretory, CD63 Tetraspanin Tag) = 10ug
SDP
Catalog No. NC3244669
Encompass_Preferred
Label exosomes with GFP by expressing the pCT-CD63-GFP Cyto-Tracer in the parent cells. Great for tracking exosomes and monitoring cargo delivery. Monitor exosome dynamics with the pCT-CD63-GFP (pCMV) Exosome Cyto-Tracer (also available as a CD63-RFP fusion, pCT-CD63-RFP). Provided as either lentivector or pre-packaged virus, this Cyto-Tracer fuses GFP to the tetraspanin CD63, which then gets packaged into exosomes produced by the transfected cells. Great for creating stable cell lines that produce GFP-labeled exosomes as well as labeling exosomes from primary cells, tumor cells, stem cells, and more.
Catalog No. NC3244669 Supplier System Biosciences LLC Supplier No. CYTO120PA1
Label exosomes with GFP by expressing the pCT-CD63-GFP Cyto-Tracer in the parent cells. Great for tracking exosomes and monitoring cargo delivery. Monitor exosome dynamics with the pCT-CD63-GFP (pCMV) Exosome Cyto-Tracer (also available as a CD63-RFP fusion, pCT-CD63-RFP). Provided as either lentivector or pre-packaged virus, this Cyto-Tracer fuses GFP to the tetraspanin CD63, which then gets packaged into exosomes produced by the transfected cells. Great for creating stable cell lines that produce GFP-labeled exosomes as well as labeling exosomes from primary cells, tumor cells, stem cells, and more.