Learn More
Zen Bio Inc PCT EPI KERATIN MED LOW BPE
SDP
Catalog No. NC9604824
Encompass
Low-BPE, PCT medium recommended where absolute best attachement, isolation efficiency, colony formation efficiency and longevity is required, the quality of the starting material is not optimal, and / or a low BPE concentration (5 to 8 x lower than competitors) is compatible with the experimental setup.
Catalog No. NC9604824 Supplier Zen Bio Inc Supplier No. CNT57
Low-BPE, PCT medium recommended where absolute best attachement, isolation efficiency, colony formation efficiency and longevity is required, the quality of the starting material is not optimal, and / or a low BPE concentration (5 to 8 x lower than competitors) is compatible with the experimental setup.