Learn More
Zen Bio Inc PCT EPIDERMAL KERATINOCYTE MED
SDP
Catalog No. NC9474150
Encompass
Defined, PCT media. Where a completely defined environment and maximum keratinocyte purity is required. CnT-07 provides excellent results through maximum retention of keratinocyte progenitors, and inhibition of fibroblasts
Catalog No. NC9474150 Supplier Zen Bio Inc Supplier No. CNT07
Defined, PCT media. Where a completely defined environment and maximum keratinocyte purity is required. CnT-07 provides excellent results through maximum retention of keratinocyte progenitors, and inhibition of fibroblasts