Learn More
Hampton Research HR2-940-A2/PCT #A2, 185 ml - 0.1 M Tris hydrochloride pH 8.5, 0.2 M Magnesium chloride hexahydrate, 30% Polyethylene glycol 4,000
SDP
Catalog No. 502102
Encompass
HR2-940-A2/PCT #A2, 185 ml - 0.1 M Tris hydrochloride pH 8.5, 0.2 M Magnesium chloride hexahydrate, 30% Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-210-2 Supplier Hampton Research Supplier No. HR2940A2
HR2-940-A2/PCT #A2, 185 ml - 0.1 M Tris hydrochloride pH 8.5, 0.2 M Magnesium chloride hexahydrate, 30% Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.