Learn More
Hampton Research HR2-940-B2/PCT #B2, 185 ml - 0.1 M Tris hydrochloride pH 8.5, 0.2 M Magnesium chloride hexahydrate, 15% Polyethylene glycol 4,000
SDP
Catalog No. 502104
Encompass
HR2-940-B2/PCT #B2, 185 ml - 0.1 M Tris hydrochloride pH 8.5, 0.2 M Magnesium chloride hexahydrate, 15% Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-210-4 Supplier Hampton Research Supplier No. HR2940B2
HR2-940-B2/PCT #B2, 185 ml - 0.1 M Tris hydrochloride pH 8.5, 0.2 M Magnesium chloride hexahydrate, 15% Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.