Learn More
Sterlitech Corporation Polycarbonate (PCTE) Membrane Filters, 0.4 Micron, 25mm, 100/Pk
SDP
Catalog No. 501952371
Encompass_Preferred
Polycarbonate Track Etch (PCTE) membranes are made from a thin, microporous polycarbonate film material. It is ideally suited for use in blood assays and high-purity and general filtration.
Catalog No. 50-195-2371 Supplier Sterlitech Corporation Supplier No. PCT0425100
Add to Cart
Add to Cart
Polycarbonate Track Etch (PCTE) membranes are made from a thin, microporous polycarbonate film material. It is ideally suited for use in blood assays and high-purity and general filtration.