Learn More
Sterlitech Corporation Polycarbonate (PCTE) Membrane Filters, PVP-Free, 1.0 Micron, 25mm, 100/Pk
SDP
Catalog No. NC2943919
Encompass_Preferred
Polycarbonate (PCTE) Membrane Filters, PVP-Free, 1.0 Micron, 25mm, 100/PkPrecisely controlled cylindrical pores for maximum absolute particulate captureTransparent, smooth flat surface for microscopic workLowest, non-specific binding membraneNarrow pore size distributionNo sloughing or particle shedding, one integral plastic filmNegligible adsorption or absorptionLow extractables
Catalog No. NC2943919 Supplier Sterlitech Corporation Supplier No. PCTF1025100
Polycarbonate (PCTE) Membrane Filters, PVP-Free, 1.0 Micron, 25mm, 100/PkPrecisely controlled cylindrical pores for maximum absolute particulate captureTransparent, smooth flat surface for microscopic workLowest, non-specific binding membraneNarrow pore size distributionNo sloughing or particle shedding, one integral plastic filmNegligible adsorption or absorptionLow extractables