Learn More
Sterlitech Corporation Polycarbonate (PCTG) Gold-Coated Membrane Filters, 0.4 Micron, 40/20nm Coating, 25mm, 10/Pk
SDP
Catalog No. 501952372
Encompass_Preferred
Polycarbonate track-etched membranes feature a plain surface and round pores, providing ideal properties for particle analysis.Applications:* Asbestos testing* SER-s based detection for pahtogen concentration* Sample Preparation* Low protein binding* Smooth flat surface for good visibility of particles
Catalog No. 50-195-2372 Supplier Sterlitech Corporation Supplier No. 1270004
Add to Cart
Add to Cart
Polycarbonate track-etched membranes feature a plain surface and round pores, providing ideal properties for particle analysis.Applications:* Asbestos testing* SER-s based detection for pahtogen concentration* Sample Preparation* Low protein binding* Smooth flat surface for good visibility of particles