Learn More
Sino Biological Recombinant Human PCTK1 (CDK16) & Cyclin Y Protein (GST Tag), Active
Catalog No. 502573906
Encompass_Preferred
Recombinant human PCTK1 (CDK16) (107-end) and Cyclin Y (2-end) were co-expressed by baculovirus in Sf9 insect cells using N-terminal GST tags.
Catalog No. 50-257-3906 Supplier Sino Biological Supplier No. P0911G500
Recombinant human PCTK1 (CDK16) (107-end) and Cyclin Y (2-end) were co-expressed by baculovirus in Sf9 insect cells using N-terminal GST tags.