Learn More
Aviva Systems Biology Corp PCTP Antibody - middle region (ARP64248_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64248_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PCTP. It was validated on Western Blot. . Alternate names: STARD2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-214