Learn More
Aviva Systems Biology Corp PCYOX1 Antibody - C-terminal region (ARP33007_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33007_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PCYOX1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human PCYOX1. Alternate names: PCL1ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit, Yeast
Catalog No. 50-014-54850