Learn More
Aviva Systems Biology Corp PCYT2 Antibody - middle region (ARP43577_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43577_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PCYT2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PCYT2. Alternate names: ETApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-58577