Learn More
ABclonal Technology PCYT2 Rabbit pAb
SDP
Catalog No. 5000105311
Encompass_Preferred
This gene encodes an enzyme that catalyzes the formation of CDP-ethanolamine from CTP and phosphoethanolamine in the Kennedy pathway of phospholipid synthesis. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-05311 Supplier ABclonal Technology Supplier No. A153091000UL
This gene encodes an enzyme that catalyzes the formation of CDP-ethanolamine from CTP and phosphoethanolamine in the Kennedy pathway of phospholipid synthesis. Alternative splicing results in multiple transcript variants.