Learn More
AdipoGen PD 0325901 (1 mg)
SDP
Supplier:  AdipoGen AGCR13711M001
Encompass_Preferred
Chemical. CAS: 391210-10-9. Formula: C16H14F3IN2O4. MW: 482.2. PD 0325901 is a potent and selective non-ATP competitive orally active MEK1 and MEK2 inhibitor. In comparison to the CI-1040 MEK inhibitor that PD 0325901 has a greater potency of inhibition, longer duration, greater solubility, improved bioavailability and increased metabolic stability. The dual specific threonine/tyrosine kinase MEK is a key component of the RAS/RAF/MEK/ERK signaling pathway that is frequently activated in human tumors. PD 0325901 is an anti-cancer agent that is highly effective at inhibiting cell growth and proliferation in various cancer cells. It has been shown to inhibit MEK activity and suppresses phosphorylation of ERK in mouse colon 26 cells (IC50 = 0.33nM) and the growth of melanoma cell lines in vitro and in vivo. PD 0325901 promotes reprogramming of human somatic cells to iPSCs and enhances generation and self-renewal of induced pluripotent stem cells (iPSCs) in combination with other compounds.
Catalog No. 50-205-4088