Learn More
Selleck Chemical LLC Palbociclib (PD-0332991) HCl 5mg 827022-32-2
SDP
Catalog No. 508547
Encompass_Preferred
Palbociclib (PD-0332991) HCl is a highly selective inhibitor of CDK4/6 with IC50 of 11 nM/16 nM in cell-free assays, respectively. It shows no activity against CDK1/2/5, EGFR, FGFR, PDGFR, InsR, etc. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-854-7 Supplier Selleck Chemical LLC Supplier No. S11165MG
Palbociclib (PD-0332991) HCl is a highly selective inhibitor of CDK4/6 with IC50 of 11 nM/16 nM in cell-free assays, respectively. It shows no activity against CDK1/2/5, EGFR, FGFR, PDGFR, InsR, etc. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.