Learn More
Selleck Chemical LLC Palbociclib (PD0332991) Isethionate 50mg 827022-33-3
SDP
Catalog No. 505610
Encompass_Preferred
Palbociclib (PD0332991) Isethionate is a highly selective inhibitor of CDK4/6 with IC50 of 11 nM/16 nM in cell-free assays. It shows no activity against CDK1/2/5, EGFR, FGFR, PDGFR, InsR, etc. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-561-0 Supplier Selleck Chemical LLC Supplier No. S157950MG
Palbociclib (PD0332991) Isethionate is a highly selective inhibitor of CDK4/6 with IC50 of 11 nM/16 nM in cell-free assays. It shows no activity against CDK1/2/5, EGFR, FGFR, PDGFR, InsR, etc. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.