Learn More
ACROBiosystems ACROBiosystems Rabbit PD-1 / PDCD1 Protein, Fc Tag
Supplier:  ACROBiosystems PD1RB52511MG
Encompass_Preferred
Recombinant Protein;1MG;Rabbit PD-1, Fc Tag (PD1-RB5251) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # G1SUF0).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.6 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Mouse PD-L1, His Tag (Cat. No. PD1-M5220) at 10 μg/mL (100 μL/well) can bind Rabbit PD-1, Fc Tag (Cat. No. PD1-RB5251) with a linear range of 3-50 ng/mL (QC tested).
Catalog No. 50-195-7257