Learn More
ACROBiosystems ACROBiosystems Biotinylated Mouse PD-1 / PDCD1 Protein, Fc,Avitag™
Catalog No. 501957533
Encompass_Preferred
Recombinant Protein;200UG;MABSol Biotinylated Mouse PD-1, Fc,Avitag (PD1-M82F4) is expressed from human HEK293 cells. It contains AA Leu 25 - Gln 167 (Accession # NP_032824).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 44.6 kDa. The protein migrates as 53-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse PD-L1, Fc Tag (Cat. No. PD1-M5251) at 10 μg/mL (100 μL/well) can bind Biotinylated Mouse PD-1, Fc,Avitag (Cat. No. PD1-M82F4) with a linear range of 0.078-1.25 μg/mL (QC tested).;Immobilized Mouse PD-L2, His Tag (Cat. No. PD2-M52E3) at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse PD-1, Fc,Avitag (Cat. No. PD1-M82F4) with a linear range of 0.156-2.5 μg/mL (Routinely tested).
Catalog No. 50-195-7533 Supplier ACROBiosystems Supplier No. PD1M82F4200UG
Recombinant Protein;200UG;MABSol Biotinylated Mouse PD-1, Fc,Avitag (PD1-M82F4) is expressed from human HEK293 cells. It contains AA Leu 25 - Gln 167 (Accession # NP_032824).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 44.6 kDa. The protein migrates as 53-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse PD-L1, Fc Tag (Cat. No. PD1-M5251) at 10 μg/mL (100 μL/well) can bind Biotinylated Mouse PD-1, Fc,Avitag (Cat. No. PD1-M82F4) with a linear range of 0.078-1.25 μg/mL (QC tested).;Immobilized Mouse PD-L2, His Tag (Cat. No. PD2-M52E3) at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse PD-1, Fc,Avitag (Cat. No. PD1-M82F4) with a linear range of 0.156-2.5 μg/mL (Routinely tested).