Learn More
Medchemexpress LLC HY-P7395 50ug Medchemexpress, PD-1, Fc Chimera (CHO-expressed), Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP739550UG
Encompass_Preferred
Medchemexpress, HY-P7395 50ug PD-1, Fc Chimera (CHO-expressed), Human CAS: Recombinant Human PD-1, Fc Chimera (CHO-expressed) suppress activating signals from the T cell receptor when bound by either of its ligands, programmed death-ligand 1 (PD-L1) or PD-L2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3157