Learn More
ACROBiosystems ACROBiosystems Human PD-1 / PDCD1 Protein, His Tag, low endotoxin (HPLC-verified)
Supplier:  ACROBiosystems PD1H522A100UG
Encompass_Preferred
Recombinant Protein;100UG;Human PD-1, His Tag, low endotoxin (HPLC-verified) (PD1-H522a) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # NP_005009.2).;This protein carries a polyhistidine tag at the C-terminus, and has a calculated MW of 16.8 kDa. The N-terminus Sequence Analysis is Leu 25. The reducing (R) protein migrates as 31-44 kDa in SDS-PAGE due to glycosylation.;Immobilized Human PD-1, His Tag, low endotoxin (HPLC-verified) (Cat. No. PD1-H522a) at 2 μg/mL (100 μL/well) can bind Human PD-L2, Mouse IgG1 Fc Tag (Cat. No. PD2-H52A5) with a linear range of 10-156 ng/mL (Routinely tested).
Catalog No. 50-195-7190