Learn More
ACROBiosystems ACROBiosystems Rat PD-1 / PDCD1 Protein, His Tag
Catalog No. 501957283
Encompass_Preferred
Recombinant Protein;100UG;Rat PD-1, His Tag (PD1-R52H2) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # NP_001100397.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.1 kDa. The protein migrates as 30-44 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Rat PD-1, His Tag (Cat. No. PD1-R52H2) at 2 μg/mL (100 μL/well) can bind Mouse PD-L1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. PD1-M52A2) with a linear range of 0.02-0.313 μg/mL (QC tested).
Catalog No. 50-195-7283 Supplier ACROBiosystems Supplier No. PD1R52H2100UG
Recombinant Protein;100UG;Rat PD-1, His Tag (PD1-R52H2) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # NP_001100397.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.1 kDa. The protein migrates as 30-44 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Rat PD-1, His Tag (Cat. No. PD1-R52H2) at 2 μg/mL (100 μL/well) can bind Mouse PD-L1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. PD1-M52A2) with a linear range of 0.02-0.313 μg/mL (QC tested).