Learn More
ACROBiosystems ACROBiosystems Biotinylated Cynomolgus PD-1 / PDCD1 Protein, His,Avitag™
Catalog No. 501956508
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Cynomolgus PD-1, His,Avitag (PD1-C82E6) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # B0LAJ3).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 19.5 kDa. The protein migrates as 30-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Cynomolgus PD-1, His,Avitag (Cat. No. PD1-C82E6) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Cynomolgus / Rhesus macaque PD-L1, Fc Tag (Cat. No. PD1-C5253) with a linear range of 0.2-5 ng/mL (QC tested).;Immobilized Biotinylated Cynomolgus PD-1, His,Avitag (Cat. No. PD1-C82E6) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Nivolumab with a linear range of 1-20 ng/mL (Routinely tested).
Catalog No. 50-195-6508 Supplier ACROBiosystems Supplier No. PD1C82E6200UG
Recombinant Protein;200UG;Biotinylated Cynomolgus PD-1, His,Avitag (PD1-C82E6) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # B0LAJ3).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 19.5 kDa. The protein migrates as 30-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Cynomolgus PD-1, His,Avitag (Cat. No. PD1-C82E6) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Cynomolgus / Rhesus macaque PD-L1, Fc Tag (Cat. No. PD1-C5253) with a linear range of 0.2-5 ng/mL (QC tested).;Immobilized Biotinylated Cynomolgus PD-1, His,Avitag (Cat. No. PD1-C82E6) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Nivolumab with a linear range of 1-20 ng/mL (Routinely tested).