Learn More
BPS Bioscience Inc PD-1 (Human), Fc Fusion (Mouse IgG2a) HiP™ | 100ug
SDP
Supplier:  BPS Bioscience Inc 71997
Encompass
Human secreted programmed cell death 1 (PD-1)-Fc fusion protein, also known as, PDCD1, SLEB2, CD279 and HPD-L, GenBank Accession No. NM_005018, a.a 25-167, fused at the C-terminus to the Fc portion of Mouse IgG2a, expressed in a HEK293 cell expression system. MW = 43 kDa.
Catalog No. NC1586383