Learn More
Medchemexpress LLC CA-170 | 1673534-76-3 | 5mg
SDP
Catalog No. 501932034
Encompass_Preferred
CA-170 is an orally delivered dual inhibitor of VISTA and PD-L1. CA-170 exhibits potent rescue of proliferation and effector functions of T cells inhibited by PD-L1/L2 and VISTA with selectivity over other immune checkpoint proteins as well as a broad panel of receptors and enzymes. Purity: 99.63%. For research use only. Does not sell to patients.
Catalog No. 50-193-2034 Supplier Medchemexpress LLC Supplier No. HY1010935MG
CA-170 is an orally delivered dual inhibitor of VISTA and PD-L1. CA-170 exhibits potent rescue of proliferation and effector functions of T cells inhibited by PD-L1/L2 and VISTA with selectivity over other immune checkpoint proteins as well as a broad panel of receptors and enzymes. Purity: 99.63%. For research use only. Does not sell to patients.