Learn More
Medchemexpress LLC HY-101097 5mg Medchemexpress, PD-1-IN-17 CAS:1673560-66-1 Purity:>98%
SDP
Catalog No. 501932037
Encompass_Preferred
Medchemexpress, HY-101097 5mg PD-1-IN-17 CAS:1673560-66-1 PD-1-IN-17 is a programmed cell death-1 ( PD-1 ) inhibitor extracted from patent WO2015033301A1, Compound 12, inhibits 92% splenocyte proliferation at 100 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2037 Supplier Medchemexpress LLC Supplier No. HY1010975MG
Medchemexpress, HY-101097 5mg PD-1-IN-17 CAS:1673560-66-1 PD-1-IN-17 is a programmed cell death-1 ( PD-1 ) inhibitor extracted from patent WO2015033301A1, Compound 12, inhibits 92% splenocyte proliferation at 100 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.