Learn More
TARGETMOL CHEMICALS INC PD-1-IN-22 5MG
SDP
Catalog No. 502581459
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PD-1-IN-22 is a potent programmed cell death-1 (PD-1)/programmed cell death-ligand 1 (PD-L1) interaction inhibitor(IC50 of 92.3 nM). purity: 98%
Catalog No. 50-258-1459 Supplier TARGETMOL CHEMICALS INC Supplier No. T123795MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PD-1-IN-22 is a potent programmed cell death-1 (PD-1)/programmed cell death-ligand 1 (PD-L1) interaction inhibitor(IC50 of 92.3 nM). purity: 98%