Learn More
Medchemexpress LLC PD-1-IN-24 (compound 1) | 2360909-50-6 | 98.1% | C27H26F3NO3 | 10MG
SDP
Catalog No. 5000261541
Encompass_Preferred
PD-1-IN-24 (compound 1) is a small-molecule, orally active PD-1 inhibitor used in preclinical immunology and oncology research. It modulates the PD-1/PD-L1 immune checkpoint pathway and has documented in vivo activity. The compound is provided as a powder or as DMSO solutions, and is supplied with analytical documentation including molecular formula C27H26F3NO3, molecular weight 469.50, and typical purity around 98.1%.
  • Orally active small-molecule PD-1 inhibitor.
  • Validated for PD-1/PD-L1 pathway modulation in preclinical models.
  • Supplied as powder and ready-to-use DMSO solution formats.
  • High supplied purity (~98.1%) for reproducible results.
  • Available in multiple sizes suitable for screening and in vivo dosing.
  • Accompanied by safety data sheet and technical datasheet documentation.
Catalog No. 50-002-61541 Supplier Medchemexpress LLC Supplier No. HY13488610MG
PD-1-IN-24 (compound 1) is a small-molecule, orally active PD-1 inhibitor used in preclinical immunology and oncology research. It modulates the PD-1/PD-L1 immune checkpoint pathway and has documented in vivo activity. The compound is provided as a powder or as DMSO solutions, and is supplied with analytical documentation including molecular formula C27H26F3NO3, molecular weight 469.50, and typical purity around 98.1%.
  • Orally active small-molecule PD-1 inhibitor.
  • Validated for PD-1/PD-L1 pathway modulation in preclinical models.
  • Supplied as powder and ready-to-use DMSO solution formats.
  • High supplied purity (~98.1%) for reproducible results.
  • Available in multiple sizes suitable for screening and in vivo dosing.
  • Accompanied by safety data sheet and technical datasheet documentation.