Learn More
BPS Bioscience Inc PD-1 Lentivirus, 2 x 500 μl, ≥10^7 TU/ml, VSV-G pseudotyped, CMV promoter, Blasticidin selection marker, HIV-based, self-inactivating (SIN) vector
SDP
Catalog No. NC3597528
Encompass
The PD-1 Lentivirus is a replication-incompetent, HIV-based, VSV-G pseudotyped lentiviral particle designed for efficient transduction of human PD-1 into various mammalian cell types. It allows the delivery of the human Programmed Death Protein 1 (PD-1) gene under the CMV promoter, along with a Blasticidin selection marker, suitable for creating stable cell lines. This product is instrumental in PD-1 biology research and the development of PD-1-targeted therapeutics.
  • VSV-G pseudotyping enables efficient transduction of nearly all mammalian cell types.
  • Provides stable expression of human PD-1 for consistent experimental results.
  • Blasticidin resistance gene facilitates selection and enrichment of transduced cells.
  • Self-inactivating (SIN) lentivector enhances safety by preventing further viral replication.
  • Ideal for investigations into PD-1 antibodies and therapeutics for cancer and immune-related diseases.
Catalog No. NC3597528 Supplier BPS Bioscience Inc Supplier No. 82348
The PD-1 Lentivirus is a replication-incompetent, HIV-based, VSV-G pseudotyped lentiviral particle designed for efficient transduction of human PD-1 into various mammalian cell types. It allows the delivery of the human Programmed Death Protein 1 (PD-1) gene under the CMV promoter, along with a Blasticidin selection marker, suitable for creating stable cell lines. This product is instrumental in PD-1 biology research and the development of PD-1-targeted therapeutics.
  • VSV-G pseudotyping enables efficient transduction of nearly all mammalian cell types.
  • Provides stable expression of human PD-1 for consistent experimental results.
  • Blasticidin resistance gene facilitates selection and enrichment of transduced cells.
  • Self-inactivating (SIN) lentivector enhances safety by preventing further viral replication.
  • Ideal for investigations into PD-1 antibodies and therapeutics for cancer and immune-related diseases.