Learn More
Enzo Life Sciences PD-1 monoclonal antibody (4F12) (100µl)
SDP
Supplier:  Enzo Life Sciences ENZABS7370100
Encompass_Preferred
Clone: 4F12. Isotype: IgG2bκ. Immunogen: Recombinant human PDCD1 (aa 21-167) purified from E. coli. Source: Purified from ascites. Species reactivity: Human. Applications: ELISA, Flow Cytometry, ICC, IF, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.4, containing 0.02% sodium azide and 10% glycerol.
Catalog No. 50-201-3278