Learn More
Medchemexpress LLC PD-1 Mouse HEK293 500ug
SDP
Catalog No. 5000185110
Encompass_Preferred
Recombinant mouse PD-1 extracellular domain fused to an Fc tag, expressed in HEK293 cells and supplied as a purified protein for immunology research. It is intended for use in assays that require a biologically relevant, mammalian-expressed PD-1 reagent, such as binding studies and receptor-ligand interaction experiments.
  • Mammalian expression supports native folding and glycosylation.
  • Fc fusion enhances stability and promotes dimerization.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Available in a 500 μg format for scaling experiments.
  • Stable when aliquoted and stored at -20°C or -80°C for long-term use.
Catalog No. 50-001-85110 Supplier Medchemexpress LLC Supplier No. HYP70639500UG
Recombinant mouse PD-1 extracellular domain fused to an Fc tag, expressed in HEK293 cells and supplied as a purified protein for immunology research. It is intended for use in assays that require a biologically relevant, mammalian-expressed PD-1 reagent, such as binding studies and receptor-ligand interaction experiments.
  • Mammalian expression supports native folding and glycosylation.
  • Fc fusion enhances stability and promotes dimerization.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Available in a 500 μg format for scaling experiments.
  • Stable when aliquoted and stored at -20°C or -80°C for long-term use.