Learn More
ACROBiosystems ACROBiosystems Human PD-1 / PDCD1 Protein, Mouse IgG2a Fc Tag (HPLC-verified)
Supplier: ACROBiosystems PD1H5255100UG
Recombinant Protein;100UG;Human PD-1, Mouse IgG2a Fc Tag (HPLC-verified) (PD1-H5255) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # Q15116-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 42.5 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PD-L1, Fc Tag (HPLC-verified) (Cat. No. PD1-H5258) at 10 μg/mL (100 μL/well) can bind Human PD-1, Mouse IgG2a Fc Tag (HPLC-verified) (Cat. No. PD1-H5255) with a linear range of 2-40 ng/mL (QC tested).;Immobilized Human PD-1, Mouse IgG2a Fc Tag (HPLC-verified) (Cat. No. PD1-H5255) at 2 μg/mL (100 μL/well) can bind Human PD-L2, Fc Tag (Cat. No. PD2-H5251) with a linear range of 0.2-4 ng/mL (Routinely tested).