Learn More
TARGETMOL CHEMICALS INC PD-1/PD-L1-IN-10 10MG
SDP
Catalog No. 502578993
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PD-1/PD-L1-IN-10 is an orally activePD-1/PD-L1inhibitor (IC50of 2.7 nM) with potent anticancer efficacy. purity: 98%
Catalog No. 50-257-8993 Supplier TARGETMOL CHEMICALS INC Supplier No. T961610MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PD-1/PD-L1-IN-10 is an orally activePD-1/PD-L1inhibitor (IC50of 2.7 nM) with potent anticancer efficacy. purity: 98%