Learn More
Medchemexpress LLC Pd-1/pd-l1-in-23 | 2597056-04-5 | 100.0% | 703.41 g/mol | C32H30BrCl2N3O6 | 5 MG
SDP
Catalog No. 5000221131
Encompass_Preferred
PD-1/PD-L1-IN-23 is a small-molecule inhibitor of the PD-1/PD-L1 immune checkpoint provided as a research reagent. Described as an orally active ester prodrug of L7, it is supplied as a solid suitable for biochemical assays and preclinical studies. CAS 2597056-04-5; molecular formula C32H30BrCl2N3O6; molecular weight 703.41 g/mol.
  • Potent small-molecule inhibitor of PD-1/PD-L1 interaction
  • Orally active prodrug with demonstrated preclinical activity
  • Designed to convert to an active metabolite in vivo
  • Solid, white to off-white form with high purity (99.98%)
  • Soluble in DMSO; available as solution and powder formats
  • Available in multiple research-scale package sizes including 5 mg
Catalog No. 50-002-21131 Supplier Medchemexpress LLC Supplier No. HY1457745MG
PD-1/PD-L1-IN-23 is a small-molecule inhibitor of the PD-1/PD-L1 immune checkpoint provided as a research reagent. Described as an orally active ester prodrug of L7, it is supplied as a solid suitable for biochemical assays and preclinical studies. CAS 2597056-04-5; molecular formula C32H30BrCl2N3O6; molecular weight 703.41 g/mol.
  • Potent small-molecule inhibitor of PD-1/PD-L1 interaction
  • Orally active prodrug with demonstrated preclinical activity
  • Designed to convert to an active metabolite in vivo
  • Solid, white to off-white form with high purity (99.98%)
  • Soluble in DMSO; available as solution and powder formats
  • Available in multiple research-scale package sizes including 5 mg