Learn More
Medchemexpress LLC PD-1/PD-L1-IN-23 | 2597056-04-5 | 100.0% | C32H30BrCl2N3O6 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY145774100MG
Encompass_Preferred
PD-1/PD-L1-IN-23 is a potent and orally active inhibitor of PD-1/PD-L1. It is an ester proagent of L7, a benzo[c]oxadiazole derivative, and has demonstrated significant antitumor effects in tumor models of syngeneic and PD-L1 humanized mice. This product is intended for research use only.
  • Potent and orally active inhibitor of PD-1/PD-L1
  • Ester proagent of L7, a benzo[c]oxadiazole derivative
  • Evaluated as a PD-L1 inhibitor
  • Demonstrates significant antitumor effects in tumor models
  • Intended for research use only
Catalog No. 50-003-54295