Learn More
Medchemexpress LLC PD-1/PD-L1-IN-23 | 2597056-04-5 | 99.9% | C32H30BrCl2N3O6 | 50 MG
SDP
Catalog No. 5000355812
Encompass_Preferred
PD-1/PD-L1-IN-23 is a potent and orally active inhibitor of PD-1/PD-L1. It is an ester proagent of L7, a benzo[c]oxadiazole derivative biologically evaluated as an inhibitor of PD-L1. It has shown significant antitumor effects in tumor models of syngeneic and PD-L1 humanized mice.
  • Potent and orally active inhibitor of PD-1/PD-L1
  • Ester proagent of L7, a benzo[c]oxadiazole derivative
  • Displays significant antitumor effects in tumor models
  • For research use only
Catalog No. 50-003-55812 Supplier Medchemexpress LLC Supplier No. HY14577450MG
PD-1/PD-L1-IN-23 is a potent and orally active inhibitor of PD-1/PD-L1. It is an ester proagent of L7, a benzo[c]oxadiazole derivative biologically evaluated as an inhibitor of PD-L1. It has shown significant antitumor effects in tumor models of syngeneic and PD-L1 humanized mice.
  • Potent and orally active inhibitor of PD-1/PD-L1
  • Ester proagent of L7, a benzo[c]oxadiazole derivative
  • Displays significant antitumor effects in tumor models
  • For research use only