Learn More
Genscript Corporation PD-1/PDCD1 hFc ChimeraBiotin
Catalog No. 5000057062
Encompass
Programmed cell death protein 1 also known as PD-1 and CD279 is a protein found on the surface of cells that has a role in regulating the immune system s response to the cells of the human body by down-regulating the immune system and promoting self tolerance by suppressing T cell inflammatory activity
Catalog No. 50-000-57062 Supplier Genscript Corporation Supplier No. Z04582500
Programmed cell death protein 1 also known as PD-1 and CD279 is a protein found on the surface of cells that has a role in regulating the immune system s response to the cells of the human body by down-regulating the immune system and promoting self tolerance by suppressing T cell inflammatory activity