Learn More
Sino Biological Recombinant Canine PD-1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 70109D08H100
Encompass_Preferred
A DNA sequence encoding the canine PDCD1 (XP_022265285.1) (Met1-Leu169) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-4511