Learn More
Sino Biological Recombinant Mouse PD-1 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 50124M08HB100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Gln167) of mouse PDCD1 (NP_032824.1) precursor was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-6197