Learn More
Enzo Life Sciences PD-1 Recombinant monoclonal antibody (RMPD1-1) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS3370100
Encompass_Preferred
Clone: RMPD1-1. Isotype: IgGκ. Immunogen: Recombinant full length human PD-1. Species reactivity: Human. Applications: IF, IHC (PS). Purity: Protein A affinity purified. Formulation: Liquid. In 1X PBS containing 0.1 mg/ml BSA (US sourced) and 0.05% sodium azide.
Catalog No. 50-201-2868