Learn More
Eurofins Discoverx PathHunter PD-1 Signaling Bioassay Kit, 2pl. (93-1104Y19-00117)
Catalog No. 502002102
Encompass
The PathHunter PD-1 Signaling Bioassay Kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This cell line enables assessment of ligand (e.g. PD-L1, PD-L2, activating antibody) based activation of PD-1 receptors via detection of SH2 domain protein recruitment. The included cell line overexpresses ProLink-tagged PD-1 target protein and EA-tagged SH2 domain protein. Upon activation of the receptor by its ligand or an agonist antibody, recruitment of SH2 domain protein to the phosphorylated receptor tail leads to production of EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.
Catalog No. 50-200-2102 Supplier Eurofins Discoverx Supplier No. 931104Y1900117
Add to Cart
Add to Cart
The PathHunter PD-1 Signaling Bioassay Kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This cell line enables assessment of ligand (e.g. PD-L1, PD-L2, activating antibody) based activation of PD-1 receptors via detection of SH2 domain protein recruitment. The included cell line overexpresses ProLink-tagged PD-1 target protein and EA-tagged SH2 domain protein. Upon activation of the receptor by its ligand or an agonist antibody, recruitment of SH2 domain protein to the phosphorylated receptor tail leads to production of EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.