Learn More
Medchemexpress LLC PD 102807 5mg | 23062-91-1 | 5MG
SDP
Catalog No. 5000241242
Encompass_Preferred
PD 102807 is a M4 muscarinic receptor antagonist with an IC50 of 90 7 nM PD 102807 inhibits M1 M2 M3 M5 muscarinic receptor with IC50s of 6558 7 3440 7 950 0 and 7411 7 nM respectively[1 Antidyskinetic effect
Catalog No. 50-002-41242 Supplier Medchemexpress LLC Supplier No. HY1076465MG
PD 102807 is a M4 muscarinic receptor antagonist with an IC50 of 90 7 nM PD 102807 inhibits M1 M2 M3 M5 muscarinic receptor with IC50s of 6558 7 3440 7 950 0 and 7411 7 nM respectively[1 Antidyskinetic effect