Learn More
Medchemexpress LLC HY-100032 5mg Medchemexpress, PD 117519 CAS:96392-15-3 Purity:>98%
SDP
Catalog No. 501931702
Encompass_Preferred
Medchemexpress, HY-100032 5mg PD 117519 CAS:96392-15-3 PD 117519 (CI947) is an A 2A adenosine agonist which has shown oral antihypertensive activity in pharmacological animal models [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1702 Supplier Medchemexpress LLC Supplier No. HY1000325MG
Medchemexpress, HY-100032 5mg PD 117519 CAS:96392-15-3 PD 117519 (CI947) is an A 2A adenosine agonist which has shown oral antihypertensive activity in pharmacological animal models [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.