Learn More
Medchemexpress LLC -PD 128907 hydroc 10mM | 300576-59-4 | 98.92% | 285.77 | 50 MG
SDP
Catalog No. 5000344718
Encompass_Preferred
(+)-PD 128907 hydrochloride is a selective dopamine D2/D3 receptor agonist. It demonstrates high selectivity for human and rat D3 receptors, with Ki values of 1.7 nM and 0.84 nM respectively, showing over 100-fold and 900-fold selectivity compared to human and rat D2 receptors. This compound is intended for research use only.
  • Selective dopamine D2/D3 receptor agonist.
  • Exhibits high affinity for D3 receptors in both human and rat models.
  • Shows significant selectivity over D2 receptors.
  • Decreases dialysate dopamine levels in wild-type mice, being more potent than in D3 knock-out mice.
Catalog No. 50-003-44718 Supplier Medchemexpress LLC Supplier No. HY11000010MM/1ML
(+)-PD 128907 hydrochloride is a selective dopamine D2/D3 receptor agonist. It demonstrates high selectivity for human and rat D3 receptors, with Ki values of 1.7 nM and 0.84 nM respectively, showing over 100-fold and 900-fold selectivity compared to human and rat D2 receptors. This compound is intended for research use only.
  • Selective dopamine D2/D3 receptor agonist.
  • Exhibits high affinity for D3 receptors in both human and rat models.
  • Shows significant selectivity over D2 receptors.
  • Decreases dialysate dopamine levels in wild-type mice, being more potent than in D3 knock-out mice.