Learn More
Medchemexpress LLC PD 128907 hydrochloride | 112960-16-4 | MFCD01529976 | 99.5% | 285.77 | C14H20ClNO3 | 25 MG
SDP
Catalog No. 5000214167
Encompass_Preferred
PD 128907 hydrochloride is a potent and selective dopamine D3 receptor agonist supplied as a solid biochemical reagent for life-science research. It is used in pharmacology and neurobiology to probe D3-mediated signaling and dopaminergic mechanisms, and is provided at high purity in small pack sizes for laboratory experiments.
  • Potent D3 receptor agonist with low-nanomolar activity.
  • High selectivity over D2 receptors for receptor-specific studies.
  • High purity suitable for research applications.
  • Solid form with white to light yellow appearance.
  • Available in multiple small pack sizes for dosing flexibility.
  • Recommended sealed storage away from moisture to preserve stability.
Catalog No. 50-002-14167 Supplier Medchemexpress LLC Supplier No. HY100539A25MG
PD 128907 hydrochloride is a potent and selective dopamine D3 receptor agonist supplied as a solid biochemical reagent for life-science research. It is used in pharmacology and neurobiology to probe D3-mediated signaling and dopaminergic mechanisms, and is provided at high purity in small pack sizes for laboratory experiments.
  • Potent D3 receptor agonist with low-nanomolar activity.
  • High selectivity over D2 receptors for receptor-specific studies.
  • High purity suitable for research applications.
  • Solid form with white to light yellow appearance.
  • Available in multiple small pack sizes for dosing flexibility.
  • Recommended sealed storage away from moisture to preserve stability.