Learn More
Medchemexpress LLC (+)-PD 128907 hydrochloride | 300576-59-4 | 99.5% | 285.77 | 25 MG
SDP
Catalog No. 5000347571
Encompass_Preferred
(+)-PD 128907 hydrochloride is a selective dopamine D2/D3 receptor agonist. It functions as a biochemical reagent and can be utilized as a biological material or organic compound for life science research. It exhibits high selectivity for dopamine D3 receptors over D2 receptors.
  • Potent and selective dopamine D3 receptor agonist.
  • Acts on D2 and D3 dopamine receptors.
  • Inhibits dialysate DA levels in D3 knockout mice.
  • Displaces [3H]spiperone binding from dopamine D3 receptors.
  • Involved in GPCR/G protein and neuronal signaling pathways.
Catalog No. 50-003-47571 Supplier Medchemexpress LLC Supplier No. 25MGHY110000
(+)-PD 128907 hydrochloride is a selective dopamine D2/D3 receptor agonist. It functions as a biochemical reagent and can be utilized as a biological material or organic compound for life science research. It exhibits high selectivity for dopamine D3 receptors over D2 receptors.
  • Potent and selective dopamine D3 receptor agonist.
  • Acts on D2 and D3 dopamine receptors.
  • Inhibits dialysate DA levels in D3 knockout mice.
  • Displaces [3H]spiperone binding from dopamine D3 receptors.
  • Involved in GPCR/G protein and neuronal signaling pathways.