Learn More
Medchemexpress LLC HY-108512 2mg Medchemexpress, PD 144418 CAS:154130-99-1 Purity:>98%
SDP
Catalog No. 501968566
Encompass_Preferred
Medchemexpress, HY-108512 2mg PD 144418 CAS:154130-99-1 PD 144418 is a highly affinity, potent and selective sigma 1 (σ1) receptor ligand (Ki values of 0.08 nM and 1377 nM for σ1 and σ2 respectively), devoid of any significant affinity for other receptors, ion channels and enzymes. PD 144418 shows potential antipsychotic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8566 Supplier Medchemexpress LLC Supplier No. HY1085122MG
Medchemexpress, HY-108512 2mg PD 144418 CAS:154130-99-1 PD 144418 is a highly affinity, potent and selective sigma 1 (σ1) receptor ligand (Ki values of 0.08 nM and 1377 nM for σ1 and σ2 respectively), devoid of any significant affinity for other receptors, ion channels and enzymes. PD 144418 shows potential antipsychotic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.