Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD 144418 oxalate, Tocris Bioscience™

Catalog No. 260610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-061-0 10 mg
26-065-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 26-061-0 Supplier R&D Systems Supplier No. 2606/10

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

High affinity, selective σ1 ligand

High affinity, selective σ1 ligand (Ki values are 0.08 and 1377 nM for σ1 and σ2 respectively). Displays no significant activity at a wide range of other receptors, ion channels and enzymes. Antagonizes mescaline-induced scratching in mice following i.p. administration. Also attenuates cocaine-induced hyperactivity in mice.

Chemical Identifiers

CAS 1794760-28-3
Molecular Formula C20H24N2O5
Molecular Weight (g/mol) 372.421
InChI Key IWSFHSVGBKPYFN-UHFFFAOYSA-N
Synonym pd 144418 oxalate, 1,2,3,6-tetrahydro-5-3-4-methylphenyl-5-isoxazolyl-1-propylpyridine oxalate, pyridine,1,2,3,6-tetrahydro-5-3-4-methylphenyl-5-isoxazolyl-1-propyl, 3-3-4-methylphenyl-1,2-oxazol-5-yl-1-propyl-5,6-dihydro-2h-pyridine; oxalic acid, oxalic acid-5-3-4-methylphenyl-1,2-oxazol-5-yl-1-propyl-1,2,3,6-tetrahydropyridine 1/1
PubChem CID 53447361
IUPAC Name 3-(4-methylphenyl)-5-(1-propyl-3,6-dihydro-2H-pyridin-5-yl)-1,2-oxazole;oxalic acid
SMILES CCCN1CCC=C(C1)C2=CC(=NO2)C3=CC=C(C=C3)C.C(=O)(C(=O)O)O

Specifications

Quantity 10 mg
Formula Weight 372.42
Percent Purity >98%
Chemical Name or Material PD 144418 oxalate
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.