Learn More
Cayman Chemical PD 146176 Inhibitors
SDP
Catalog No. 502889490
Encompass_Preferred
Inhibitor of reticulocyte 15-LO-1; limits hypercholesterolemia-induced atherosclerosis in rabbits and reduces oxidant stress-induced apoptosis in endothelial cells; inhibits amyloid β protein aggregate formation without changing total levels of APP in cells stably expressing APP
Catalog No. 50-288-9490 Supplier Cayman Chemical Supplier No. 1001051850 MG
Add to Cart
Add to Cart
Inhibitor of reticulocyte 15-LO-1; limits hypercholesterolemia-induced atherosclerosis in rabbits and reduces oxidant stress-induced apoptosis in endothelial cells; inhibits amyloid β protein aggregate formation without changing total levels of APP in cells stably expressing APP