Learn More
Abcam PD 150606, Calpain inhibitor, 5MG
Catalog No. 502706593
Encompass_Preferred
MW 306.12 Da, Purity >98%. Potent, selective and cell-permeable Ca²+-dependent calpain inhibitor (Ki values are 210 and 370 nM for calpain-1 and calpain-2 respectively). Non-competitive and non-peptide. Irreversibly inhibits ICa through AMPA receptors. Anti-ischemic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6593 Supplier Abcam Supplier No. AB1414645MG
MW 306.12 Da, Purity >98%. Potent, selective and cell-permeable Ca²+-dependent calpain inhibitor (Ki values are 210 and 370 nM for calpain-1 and calpain-2 respectively). Non-competitive and non-peptide. Irreversibly inhibits ICa through AMPA receptors. Anti-ischemic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement