Learn More
Apexbio Technology LLC PD 151746, 25mg. Cas: 179461-52-0 MFCD: MFCD03453564. calpain inhibitor, cell-permeable
SDP
Supplier:  Apexbio Technology LLC B479125
Encompass_Preferred
PD 151746 is a potent and selective inhibitor of calpain with Ki value of 0.26 M for -Calpain. Calpain is a calcium-dependent, non-lysosomal cysteine protease that expressed in mammals and many other organisms. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5361
 Restricted Item